Assay Details
| Gene Symbol: cops3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017238.2 | NP_001017238.1 | 5 - 6 | 519 | 64 | |
| GenBank mRNA | CR760202.2 | - | 5 - 6 | 519 | 64 | |
More Information
Set Membership: |
|

cops3
Genomic Map