Assay Details
| Gene Symbol: med30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017241.2 | NP_001017241.1 | 3 - 4 | 466 | 96 | |
| GenBank mRNA | BC160461.1 | - | 3 - 4 | 582 | 96 | |
| CR760181.2 | - | 3 - 4 | 466 | 96 | ||
More Information
Set Membership: |
|

med30
Genomic Map