Assay Details
| Gene Symbol: pop4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017252.2 | NP_001017252.1 | 4 - 5 | 424 | 61 | |
| GenBank mRNA | BC161032.1 | - | 5 - 6 | 576 | 61 | |
| CR760064.2 | - | 4 - 5 | 424 | 61 | ||
More Information
Set Membership: |
|

pop4
Genomic Map