Assay Details
| Gene Symbol: capza2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017254.2 | NP_001017254.1 | 1 - 2 | 40 | 65 | |
| GenBank mRNA | CR760059.2 | - | 1 - 2 | 40 | 65 | |
More Information
Set Membership: |
|

capza2
Genomic Map