Assay Details
| Gene Symbol: tbl1xr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017274.2 | NP_001017274.1 | 5 - 6 | 506 | 63 | |
| XM_012963063.2 | XP_012818517.1 | 5 - 6 | 804 | 63 | ||
| XM_012963064.2 | XP_012818518.1 | 4 - 5 | 561 | 63 | ||
| XM_018093915.1 | XP_017949404.1 | 6 - 7 | 893 | 63 | ||
| XM_018093916.1 | XP_017949405.1 | 4 - 5 | 742 | 63 | ||
| GenBank mRNA | BC123966.1 | - | 5 - 6 | 487 | 63 | |
| CR855768.2 | - | 5 - 6 | 506 | 63 | ||
More Information
Set Membership: |
|

tbl1xr1
Genomic Map