Assay Details
| Gene Symbol: psap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017285.2 | NP_001017285.1 | 6 - 7 | 908 | 64 | |
| GenBank mRNA | BC161033.1 | - | 6 - 7 | 914 | 64 | |
| CR855698.2 | - | 6 - 7 | 908 | 64 | ||
More Information
Set Membership: |
|

psap
Genomic Map