Assay Details
| Gene Symbol: polr3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017346.2 | NP_001017346.1 | 4 - 5 | 408 | 72 | |
| XM_012959121.2 | XP_012814575.1 | 4 - 5 | 451 | 72 | ||
| GenBank mRNA | BC121511.1 | - | 4 - 5 | 363 | 72 | |
| CR855444.2 | - | 4 - 5 | 408 | 72 | ||
More Information
Set Membership: |
|

polr3d
Genomic Map