Assay Details
| Gene Symbol: mmp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030331.2 | NP_001025502.1 | 5 - 6 | 862 | 57 | |
| XM_012957070.2 | XP_012812524.1 | 5 - 6 | 1031 | 57 | ||
| GenBank mRNA | CR760009.2 | - | 5 - 6 | 862 | 57 | |
More Information
Set Membership: |
|

mmp3
Genomic Map