Assay Details
| Gene Symbol: rap2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035115.1 | NP_001030287.1 | 1 | 452 | 64 | |
| GenBank mRNA | BC161519.1 | - | 1 | 511 | 64 | |
| CR761529.2 | - | 1 | 452 | 64 | ||
More Information
Set Membership: |
|

rap2b
Genomic Map