Assay Details
| Gene Symbol: rap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035116.1 | NP_001030288.1 | 1 - 2 | 416 | 93 | |
| XM_012957120.2 | XP_012812574.1 | 2 - 3 | 496 | 93 | ||
| XM_012957121.2 | XP_012812575.1 | 2 - 3 | 323 | 93 | ||
| GenBank mRNA | BC157162.1 | - | 1 - 2 | 466 | 93 | |
| CR761475.2 | - | 1 - 2 | 416 | 93 | ||
More Information
Set Membership: |
|

rap2a
Genomic Map