Assay Details
| Gene Symbol: pck2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039727.1 | NP_001034816.1 | 3 - 4 | 551 | 84 | |
| XM_012958314.2 | XP_012813768.1 | 3 - 4 | 707 | 84 | ||
| GenBank mRNA | BC063224.1 | - | 3 - 4 | 467 | 84 | |
| BC124062.1 | - | 3 - 4 | 474 | 84 | ||
| CR760579.2 | - | 3 - 4 | 551 | 84 | ||
More Information
Set Membership: |
|

pck2
Genomic Map