Assay Details
| Gene Symbol: p4hb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039731.1 | NP_001034820.1 | 2 - 3 | 356 | 72 | |
| GenBank mRNA | BC064877.1 | - | 2 - 3 | 361 | 72 | |
| BC161026.1 | - | 2 - 3 | 358 | 72 | ||
| CR848325.2 | - | 2 - 3 | 356 | 72 | ||
More Information
Set Membership: |
|

p4hb
Genomic Map