Assay Details
| Gene Symbol: nemp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039743.1 | NP_001034832.1 | 5 - 6 | 719 | 62 | |
| XM_012957067.2 | XP_012812521.1 | 5 - 6 | 610 | 62 | ||
| GenBank mRNA | BC091583.1 | - | 5 - 6 | 666 | 62 | |
| BC121559.1 | - | 5 - 6 | 768 | 62 | ||
| CR848245.2 | - | 5 - 6 | 719 | 62 | ||
More Information
Set Membership: |
|

nemp1
Genomic Map