Assay Details
| Gene Symbol: hoxb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203548.2 | NP_988879.1 | 1 - 2 | 617 | 84 | |
| GenBank mRNA | BC059757.1 | - | 1 - 2 | 615 | 84 | |
| CR760432.2 | - | 1 - 2 | 617 | 84 | ||
More Information
Set Membership: |
|

hoxb9
Genomic Map