Assay Details
| Gene Symbol: rhebl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203606.2 | NP_988937.1 | 5 - 6 | 401 | 93 | |
| GenBank mRNA | BC061397.1 | - | 5 - 6 | 424 | 93 | |
| CR761520.2 | - | 5 - 6 | 401 | 93 | ||
More Information
Set Membership: |
|

rhebl1
Genomic Map