Assay Details
| Gene Symbol: wrap73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203697.2 | NP_989028.2 | 11 - 12 | 1290 | 59 | |
| XM_012966270.2 | XP_012821724.1 | 9 - 10 | 1230 | 59 | ||
| XM_018095315.1 | XP_017950804.1 | 8 - 9 | 1144 | 59 | ||
| GenBank mRNA | CR761372.2 | - | 11 - 12 | 1290 | 59 | |
More Information
Set Membership: |
|

wrap73
Genomic Map