Assay Details
| Gene Symbol: hmgn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203732.2 | NP_989063.2 | 3 - 4 | 245 | 67 | |
| XM_018091141.1 | XP_017946630.1 | 3 - 4 | 199 | 67 | ||
| GenBank mRNA | CR760370.2 | - | 3 - 4 | 245 | 67 | |
More Information
Set Membership: |
|

hmgn1
Genomic Map