Assay Details
| Gene Symbol: sec11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005129.2 | NP_001005129.1 | 2 - 3 | 249 | 58 | |
| GenBank mRNA | BC077666.1 | - | 2 - 3 | 275 | 58 | |
| CR761256.2 | - | 2 - 3 | 249 | 58 | ||
More Information
Set Membership: |
|

sec11c
Genomic Map