Assay Details
| Gene Symbol: ak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006816.2 | NP_001006817.1 | 5 - 6 | 396 | 72 | |
| XM_012968239.2 | XP_012823693.2 | 13 - 14 | 1784 | 72 | ||
| GenBank mRNA | BC076704.1 | - | 5 - 6 | 398 | 72 | |
| CR855821.2 | - | 5 - 6 | 396 | 72 | ||
More Information
Set Membership: |
|

ak1
Genomic Map