Assay Details
| Gene Symbol: smndc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006856.2 | NP_001006857.1 | 3 - 4 | 614 | 97 | |
| GenBank mRNA | BC064260.1 | - | 3 - 4 | 611 | 97 | |
| BC076962.1 | - | 585 | 97 | |||
| CR942604.2 | - | 3 - 4 | 614 | 97 | ||
More Information
Set Membership: |
|

smndc1
Genomic Map