Assay Details
| Gene Symbol: tpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008073.3 | NP_001008074.1 | 2 - 3 | 284 | 69 | |
| NM_001205338.1 | NP_001192267.1 | 2 - 3 | 284 | 69 | ||
| GenBank mRNA | BC080968.1 | - | 2 - 3 | 209 | 69 | |
| CN097570.1 | - | 2 - 3 | 286 | 69 | ||
| CR848168.2 | - | 2 - 3 | 123 | 69 | ||
| CX750176.1 | - | 2 - 3 | 160 | 69 | ||
More Information
Set Membership: |
|

tpt1
Genomic Map