Assay Details
| Gene Symbol: arhgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008140.2 | NP_001008141.1 | 3 - 4 | 359 | 62 | |
| XM_012960963.2 | XP_012816417.1 | 3 - 4 | 331 | 62 | ||
| GenBank mRNA | BC081352.1 | - | 3 - 4 | 286 | 62 | |
| CR942351.2 | - | 3 - 4 | 359 | 62 | ||
More Information
Set Membership: |
|

arhgap1
Genomic Map