Assay Details
| Gene Symbol: hmgn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011031.2 | NP_001011031.1 | 2 - 3 | 85 | 84 | |
| GenBank mRNA | BC082823.1 | - | 2 - 3 | 99 | 84 | |
| CR760583.2 | - | 2 - 3 | 60 | 84 | ||
| CR761935.2 | - | 2 - 3 | 85 | 84 | ||
More Information
Set Membership: |
|

hmgn2
Genomic Map