Assay Details
| Gene Symbol: alg10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016192.3 | NP_001016192.2 | 2 - 3 | 232 | 71 | |
| XM_018092077.1 | XP_017947566.1 | 2 - 3 | 279 | 71 | ||
| GenBank mRNA | BC123914.1 | - | 2 - 3 | 232 | 71 | |
| CR761797.2 | - | 2 - 3 | 229 | 71 | ||
More Information
Set Membership: |
|

alg10
Genomic Map