Assay Details
| Gene Symbol: b4galt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016211.3 | NP_001016211.1 | 5 - 6 | 772 | 63 | |
| XM_012964682.1 | XP_012820136.1 | 6 - 7 | 739 | 63 | ||
| XM_018094645.1 | XP_017950134.1 | 5 - 6 | 551 | 63 | ||
| XM_018094646.1 | XP_017950135.1 | 5 - 6 | 539 | 63 | ||
| GenBank mRNA | BC122889.1 | - | 5 - 6 | 772 | 63 | |
| CR926330.2 | - | 6 - 7 | 712 | 63 | ||
More Information
Set Membership: |
|

b4galt5
Genomic Map