Assay Details
| Gene Symbol: impad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016215.3 | NP_001016215.2 | 4 - 5 | 801 | 83 | |
| GenBank mRNA | BC080464.1 | - | 4 - 5 | 801 | 83 | |
| CR926321.2 | - | 4 - 5 | 1090 | 83 | ||
More Information
Set Membership: |
|

impad1
Genomic Map