Assay Details
| Gene Symbol: foxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016300.3 | NP_001016300.2 | 7 - 8 | 1077 | 75 | |
| XM_012968383.2 | XP_012823837.1 | 6 - 7 | 2194 | 75 | ||
| XM_012968384.2 | XP_012823838.1 | 6 - 7 | 1398 | 75 | ||
| XM_012968386.2 | XP_012823840.1 | 8 - 9 | 1260 | 75 | ||
| XM_012968388.2 | XP_012823842.1 | 6 - 7 | 2191 | 75 | ||
| XM_012968389.2 | XP_012823843.1 | 5 - 6 | 1779 | 75 | ||
| XM_012968390.2 | XP_012823844.1 | 5 - 6 | 1330 | 75 | ||
| GenBank mRNA | BC127344.1 | - | 7 - 8 | 1077 | 75 | |
| BC160425.1 | - | 5 - 6 | 993 | 75 | ||
| CR761524.2 | - | 5 - 6 | 890 | 75 | ||
More Information
Set Membership: |
|

foxn3
Genomic Map