Assay Details
Gene Symbol: smim8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016334.3 | NP_001016334.1 | 2 - 3 | 458 | 95 | |
XM_012962981.2 | XP_012818435.1 | 2 - 3 | 384 | 95 | ||
XM_012962982.2 | XP_012818436.1 | 2 - 3 | 258 | 95 | ||
XM_012962983.1 | XP_012818437.1 | 2 - 3 | 250 | 95 | ||
XM_012962984.2 | XP_012818438.1 | 2 - 3 | 256 | 95 | ||
XM_012962985.2 | XP_012818439.1 | 2 - 3 | 263 | 95 | ||
XM_012962986.2 | XP_012818440.1 | 2 - 3 | 188 | 95 | ||
XM_012962987.2 | XP_012818441.1 | 2 - 3 | 219 | 95 | ||
XM_012962988.2 | XP_012818442.1 | 2 - 3 | 247 | 95 | ||
XM_012962989.2 | XP_012818443.1 | 2 - 3 | 188 | 95 | ||
XM_012962990.2 | XP_012818444.1 | 2 - 3 | 311 | 95 | ||
XM_012962991.2 | XP_012818445.1 | 2 - 3 | 361 | 95 | ||
XM_012962992.2 | XP_012818446.1 | 2 - 3 | 187 | 95 | ||
XM_018093882.1 | XP_017949371.1 | 2 - 3 | 316 | 95 | ||
GenBank mRNA | BC121618.1 | - | 2 - 3 | 458 | 95 | |
CR761409.2 | - | 1 - 2 | 169 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
