Assay Details
| Gene Symbol: dglucy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016461.3 | NP_001016461.2 | 10 - 11 | 1630 | 78 | |
| GenBank mRNA | BC123974.1 | - | 10 - 11 | 1630 | 78 | |
| CR760948.2 | - | 11 - 12 | 1676 | 78 | ||
More Information
Set Membership: |
|

dglucy
Genomic Map