Assay Details
| Gene Symbol: med8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016614.3 | NP_001016614.1 | 5 - 6 | 550 | 88 | |
| XM_018093055.1 | XP_017948544.1 | 5 - 6 | 592 | 88 | ||
| GenBank mRNA | BC124562.1 | - | 5 - 6 | 550 | 88 | |
| CR760516.2 | - | 5 - 6 | 535 | 88 | ||
More Information
Set Membership: |
|

med8
Genomic Map