Assay Details
Gene Symbol: retreg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016672.3 | NP_001016672.2 | 2 - 3 | 522 | 76 | |
XM_012954518.2 | XP_012809972.1 | 3 - 4 | 489 | 76 | ||
GenBank mRNA | BC121834.1 | - | 2 - 3 | 522 | 76 | |
CR760241.2 | - | 2 - 3 | 372 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
