Assay Details
| Gene Symbol: retreg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016672.3 | NP_001016672.2 | 5 - 6 | 756 | 68 | |
| XM_012954518.2 | XP_012809972.1 | 6 - 7 | 723 | 68 | ||
| XM_012954519.2 | XP_012809973.1 | 6 - 7 | 720 | 68 | ||
| XM_018097745.1 | XP_017953234.1 | 5 - 6 | 696 | 68 | ||
| GenBank mRNA | BC121834.1 | - | 5 - 6 | 756 | 68 | |
| CR760241.2 | - | 5 - 6 | 606 | 68 | ||
More Information
Set Membership: |
|

retreg3
Genomic Map