Assay Details
| Gene Symbol: eif1ad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016696.3 | NP_001016696.1 | 1 - 2 | 296 | 77 | |
| GenBank mRNA | BC121518.1 | - | 1 - 2 | 296 | 77 | |
| CR760166.2 | - | 1 - 2 | 183 | 77 | ||
More Information
Set Membership: |
|

eif1ad
Genomic Map