Assay Details
| Gene Symbol: etv6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016880.3 | NP_001016880.2 | 1196 | 74 | ||
| XM_018090091.1 | XP_017945580.1 | 5 - 6 | 1046 | 74 | ||
| GenBank mRNA | BC123025.1 | - | 1196 | 74 | ||
| CR848590.2 | - | 1046 | 74 | |||
More Information
Set Membership: |
|

etv6
Genomic Map