Assay Details
| Gene Symbol: zfand2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016899.4 | NP_001016899.2 | 7 - 8 | 826 | 67 | |
| XM_018097116.1 | XP_017952605.1 | 7 - 8 | 789 | 67 | ||
| XM_018097117.1 | XP_017952606.1 | 6 - 7 | 745 | 67 | ||
| GenBank mRNA | CR942530.2 | - | 7 - 8 | 768 | 67 | |
| DT389061.1 | - | 6 - 7 | 596 | 67 | ||
More Information
Set Membership: |
|

zfand2a
Genomic Map