Assay Details
| Gene Symbol: zbtb18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016948.3 | NP_001016948.2 | 1 | 1356 | 63 | |
| XM_012963043.2 | XP_012818497.1 | 2 | 4952 | 63 | ||
| XM_012963044.2 | XP_012818498.1 | 2 | 2776 | 63 | ||
| XM_012963045.2 | XP_012818499.1 | 2 | 4935 | 63 | ||
| XM_012963046.2 | XP_012818500.1 | 2 | 2991 | 63 | ||
| XM_012963047.2 | XP_012818501.1 | 2 | 2548 | 63 | ||
| XM_018093905.1 | XP_017949394.1 | 2 | 4772 | 63 | ||
| GenBank mRNA | BC121215.1 | - | 1 | 1356 | 63 | |
More Information
Set Membership: |
|

zbtb18
Genomic Map