Assay Details
| Gene Symbol: impa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017057.3 | NP_001017057.1 | 4 - 5 | 371 | 64 | |
| XM_012964768.1 | XP_012820222.1 | 5 - 6 | 445 | 64 | ||
| XM_012964769.1 | XP_012820223.1 | 5 - 6 | 380 | 64 | ||
| XM_018094671.1 | XP_017950160.1 | 4 - 5 | 306 | 64 | ||
| GenBank mRNA | BC121539.1 | - | 4 - 5 | 371 | 64 | |
| CR761717.2 | - | 5 - 6 | 415 | 64 | ||
More Information
Set Membership: |
|

impa1
Genomic Map