Assay Details
| Gene Symbol: fam122b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017288.3 | NP_001017288.1 | 3 - 4 | 450 | 69 | |
| GenBank mRNA | BC121207.1 | - | 3 - 4 | 450 | 69 | |
| CR855692.2 | - | 3 - 4 | 249 | 69 | ||
More Information
Set Membership: |
|

fam122b
Genomic Map