Assay Details
| Gene Symbol: cs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017313.3 | NP_001017313.1 | 1 - 2 | 338 | 56 | |
| GenBank mRNA | BC080507.1 | - | 1 - 2 | 154 | 56 | |
| BC127380.1 | - | 1 - 2 | 338 | 56 | ||
| CR855469.2 | - | 1 - 2 | 121 | 56 | ||
More Information
Set Membership: |
|

cs
Genomic Map