Assay Details
| Gene Symbol: b3gat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030353.2 | NP_001025524.1 | 1 - 2 | 969 | 66 | |
| GenBank mRNA | AJ890843.1 | - | 1 - 2 | 616 | 66 | |
| BC121201.1 | - | 1 - 2 | 969 | 66 | ||
More Information
Set Membership: |
|

b3gat2
Genomic Map