Assay Details
| Gene Symbol: ttc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043347.1 | NP_001036812.1 | 8 - 9 | 1083 | 61 | |
| GenBank mRNA | BC087820.1 | - | 8 - 9 | 1019 | 61 | |
| BC121840.1 | - | 8 - 9 | 1083 | 61 | ||
| CR942510.2 | - | 8 - 9 | 1059 | 61 | ||
More Information
Set Membership: |
|

ttc5
Genomic Map