Assay Details
| Gene Symbol: ggct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044400.1 | NP_001037865.1 | 1 - 2 | 171 | 67 | |
| XM_012964807.2 | XP_012820261.1 | 2 - 3 | 352 | 67 | ||
| GenBank mRNA | BC096512.1 | - | 1 - 2 | 132 | 67 | |
| BC121204.1 | - | 1 - 2 | 142 | 67 | ||
| BC161542.1 | - | 1 - 2 | 151 | 67 | ||
| CR942442.2 | - | 1 - 2 | 171 | 67 | ||
More Information
Set Membership: |
|

ggct
Genomic Map