Assay Details
| Gene Symbol: mcrip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044420.1 | NP_001037885.1 | 4 - 5 | 408 | 97 | |
| XM_012970504.2 | XP_012825958.1 | 4 - 5 | 478 | 97 | ||
| GenBank mRNA | BC159062.1 | - | 4 - 5 | 428 | 97 | |
| CR760130.2 | - | 4 - 5 | 408 | 97 | ||
More Information
Set Membership: |
|

mcrip2
Genomic Map