Assay Details
| Gene Symbol: hpcal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044428.1 | NP_001037893.1 | 865 | 101 | ||
| GenBank mRNA | CR760317.2 | - | 865 | 101 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hpcal1| Gene Symbol: hpcal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044428.1 | NP_001037893.1 | 865 | 101 | ||
| GenBank mRNA | CR760317.2 | - | 865 | 101 | ||
Set Membership: |
|