Assay Details
| Gene Symbol: ahcy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044429.1 | NP_001037894.1 | 9 - 10 | 1185 | 82 | |
| GenBank mRNA | BC135905.1 | - | 9 - 10 | 1192 | 82 | |
| CR760385.2 | - | 9 - 10 | 1185 | 82 | ||
More Information
Set Membership: |
|

ahcy
Genomic Map