Assay Details
| Gene Symbol: uqcr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044430.1 | NP_001037895.1 | 1 - 2 | 194 | 70 | |
| GenBank mRNA | BC155048.1 | - | 1 - 2 | 144 | 70 | |
| CR760464.2 | - | 1 - 2 | 194 | 70 | ||
More Information
Set Membership: |
|

uqcr10
Genomic Map