Assay Details
| Gene Symbol: rps19bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044432.1 | NP_001037897.1 | 3 - 4 | 335 | 86 | |
| GenBank mRNA | BC155375.1 | - | 3 - 4 | 340 | 86 | |
| CR760483.2 | - | 3 - 4 | 335 | 86 | ||
More Information
Set Membership: |
|

rps19bp1
Genomic Map