Assay Details
Gene Symbol: arl5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044444.1 | NP_001037909.1 | 2 - 3 | 144 | 80 | |
GenBank mRNA | CR760784.2 | - | 2 - 3 | 144 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
