Assay Details
| Gene Symbol: c3orf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044448.1 | NP_001037913.1 | 2 - 3 | 391 | 67 | |
| XM_018091317.1 | XP_017946806.1 | 2 - 3 | 267 | 67 | ||
| GenBank mRNA | BC160453.1 | - | 3 - 4 | 376 | 67 | |
| CR760904.2 | - | 2 - 3 | 391 | 67 | ||
More Information
Set Membership: |
|

c3orf38
Genomic Map