Assay Details
| Gene Symbol: hpca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044471.1 | NP_001037936.1 | 3 - 4 | 689 | 88 | |
| XM_012957127.2 | XP_012812581.1 | 3 - 4 | 636 | 88 | ||
| GenBank mRNA | BC135480.1 | - | 3 - 4 | 698 | 88 | |
| CR761366.2 | - | 3 - 4 | 689 | 88 | ||
More Information
Set Membership: |
|

hpca
Genomic Map